Please login to your online account to display your discounted pricing

2-Amino-2-phenylbutyric acid, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC358580050

 View more versions of this product

Catalog No. AC358580050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Quantity 5g
Color White
Beilstein 14, 512
Chemical Name or Material 2-Amino-2-phenylbutyric acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H13NO2
MDL Number MFCD00038162
CAS 5438-07-3
Synonym 2-Ethyl-2-phenylglycine
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 179.22

Specifications

Chemical Name or Material 2-Amino-2-phenylbutyric acid
Name Note 99%
CAS 5438-07-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00038162
Synonym 2-Ethyl-2-phenylglycine