Please login to your online account to display your discounted pricing

2-Amino-2-phenylbutyric acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-2-phenylbutyric acid
Name Note 99%
CAS 5438-07-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00038162
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC358580050

  SDS
thermo fisher scientific
AC358580050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Quantity 5g
Color White
Beilstein 14, 512
Chemical Name or Material 2-Amino-2-phenylbutyric acid
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H13NO2
MDL Number MFCD00038162
CAS 5438-07-3
Synonym 2-Ethyl-2-phenylglycine
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 179.22