missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-2'-O-methyladenosine, 99%

$42.86 - $153.12

Chemical Identifiers

CAS 80791-87-3
Molecular Formula C11H16N6O4
Molecular Weight (g/mol) 296.287
MDL Number MFCD01630959
InChI Key JLWUWXCKSOIFPS-KQYNXXCUSA-N
Synonym 2-amino-2'-o-methyladenosine, 2'-o-methyl-2,6-diaminopurine-riboside, 2-amino-2'-o-methyl-adenosine, 2r,3r,4r,5r-5-2,6-diaminopurin-9-yl-2-hydroxymethyl-4-methoxyoxolan-3-ol, 2'-o-methyl-2-aminoadenosine, 2-nh2-2'-o-me-a, adenosine, 2-amino-2'-o-methyl, 2?-o-methyl-2,6-diaminopurine-riboside, 2,6-diamino-9-2-o-methyl-beta-d-ribofuranosyl purine, 2,6-diamino-9-2'-o-methyl-beta-d-ribofuranosyl purine
PubChem CID 9900805
IUPAC Name (2R,3R,4R,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
SMILES COC1C(C(OC1N2C=NC3=C2N=C(N=C3N)N)CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65334MD View Documents Alfa Aesar™
J65334MD
250mg Each for $42.86
 
AAJ6533403 View Documents Alfa Aesar™
J6533403
1g Each for $153.12
 
Specifications

Chemical Identifiers

80791-87-3
296.287
JLWUWXCKSOIFPS-KQYNXXCUSA-N
9900805
COC1C(C(OC1N2C=NC3=C2N=C(N=C3N)N)CO)O
C11H16N6O4
MFCD01630959
2-amino-2'-o-methyladenosine, 2'-o-methyl-2,6-diaminopurine-riboside, 2-amino-2'-o-methyl-adenosine, 2r,3r,4r,5r-5-2,6-diaminopurin-9-yl-2-hydroxymethyl-4-methoxyoxolan-3-ol, 2'-o-methyl-2-aminoadenosine, 2-nh2-2'-o-me-a, adenosine, 2-amino-2'-o-methyl, 2?-o-methyl-2,6-diaminopurine-riboside, 2,6-diamino-9-2-o-methyl-beta-d-ribofuranosyl purine, 2,6-diamino-9-2'-o-methyl-beta-d-ribofuranosyl purine
(2R,3R,4R,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol

Specifications

White
80791-87-3
MFCD01630959
JLWUWXCKSOIFPS-KQYNXXCUSA-N
(2R,3R,4R,5R)-5-(2,6-diaminopurin-9-yl)-2-(hydroxymethyl)-4-methoxyoxolan-3-ol
9900805
Powder
2-Amino-2'-O-methyladenosine
250mg
C11H16N6O4
2-amino-2'-o-methyladenosine, 2'-o-methyl-2,6-diaminopurine-riboside, 2-amino-2'-o-methyl-adenosine, 2r,3r,4r,5r-5-2,6-diaminopurin-9-yl-2-hydroxymethyl-4-methoxyoxolan-3-ol, 2'-o-methyl-2-aminoadenosine, 2-nh2-2'-o-me-a, adenosine, 2-amino-2'-o-methyl, 2?-o-methyl-2,6-diaminopurine-riboside, 2,6-diamino-9-2-o-methyl-beta-d-ribofuranosyl purine, 2,6-diamino-9-2'-o-methyl-beta-d-ribofuranosyl purine
COC1C(C(OC1N2C=NC3=C2N=C(N=C3N)N)CO)O
296.287
296.28
99%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C