missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1,3-propanediol 98.0+%, TCI America™

Supplier:  TCI America A0332500G

 View more versions of this product

Catalog No. A0332500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Amino-2-methyl-1,3-propanediol
108°C
152°C
MFCD00004678
2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol
CC(CO)(CO)N
105.137
CHEBI:991
≥98.0% (GC,T)
115-69-5
White-Yellow
C4H11NO2
500 g
UXFQFBNBSPQBJW-UHFFFAOYSA-N
2-amino-2-methylpropane-1,3-diol
1531
105.14
Crystalline Powder

Chemical Identifiers

115-69-5
105.137
UXFQFBNBSPQBJW-UHFFFAOYSA-N
1531
2-amino-2-methylpropane-1,3-diol
C4H11NO2
MFCD00004678
2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol
CHEBI:991
CC(CO)(CO)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0321

RTECSNumber : TY2975000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions