missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-2-methyl-1,3-propanediol 99%, ACROS Organics™

Manufacturer:  Acros Organics AC104050025

 View more versions of this product

Catalog No. AC104050025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-2-methyl-1, 3-propanediol
Plastic Bottle
151.0°C (10.0 mmHg)
108.0°C to 110.0°C
99%
98.5
99%
HOCH2C(CH3)(NH2)CH2OH
04, 303
15, 445
UXFQFBNBSPQBJW-UHFFFAOYSA-N
2-amino-2-methylpropane-1,3-diol
1531
105.14
99%
Authentic
0.5% max.
White
2.5kg
115-69-5
100.0
C4H11NO2
MFCD00004678
02,24
1,1-di hydroxymethyl ethylamine
CC(CO)(CO)N
105.137
CHEBI:991
Crystalline Powder

Chemical Identifiers

115-69-5
105.137
UXFQFBNBSPQBJW-UHFFFAOYSA-N
1531
2-amino-2-methylpropane-1,3-diol
C4H11NO2
MFCD00004678
1,1-di hydroxymethyl ethylamine
CHEBI:991
CC(CO)(CO)N
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 204-100-7

RTECSNumber : TY2975000

TSCA : TSCA

SDS Documents