missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-2'-fluoro-2'-deoxyadenosine, 99%

$125.05 - $453.38

Chemical Identifiers

CAS 134444-47-6
Molecular Formula C10H13FN6O3
Molecular Weight (g/mol) 284.251
MDL Number MFCD00900994
InChI Key MHWHYOFBOWTAHZ-DXTOWSMRSA-N
Synonym 2-amino-2'-deoxy-2'-fluoroadenosine, 2'-fluoro-2,6-diaminopurine-2'-deoxyriboside, 2-amino-2'-fluoro-2'-deoxyadenosine, 2'-amino-2'-deoxy-2-fluoroadenosine, 2r,3r,4r,5r-5-2,6-diaminopurin-9-yl-4-fluoro-2-hydroxymethyl oxolan-3-ol, 2-amino-2/'-deoxy-2/'-fluoroadenosine, adenosine, 2-amino-2'-deoxy-2'-fluoro, 2,6-diamino-9-2-deoxy-2-fluoro-beta-d-ribofuranosyl-9h-purine, 1,2-dideoxy-1-2,6-diamino-9h-purin-9-yl-2-fluoro-beta-d-ribofuranose
PubChem CID 9971101
IUPAC Name (2R,3R,4R,5R)-5-(2,6-diaminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
SMILES C1=NC2=C(N1C3C(C(C(O3)CO)O)F)N=C(N=C2N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65741MD View Documents Alfa Aesar™
J65741MD
250mg Each for $125.05
 
AAJ6574103 View Documents Alfa Aesar™
J6574103
1g Each for $453.38
 
Specifications

Chemical Identifiers

134444-47-6
284.251
MHWHYOFBOWTAHZ-DXTOWSMRSA-N
9971101
C1=NC2=C(N1C3C(C(C(O3)CO)O)F)N=C(N=C2N)N
C10H13FN6O3
MFCD00900994
2-amino-2'-deoxy-2'-fluoroadenosine, 2'-fluoro-2,6-diaminopurine-2'-deoxyriboside, 2-amino-2'-fluoro-2'-deoxyadenosine, 2'-amino-2'-deoxy-2-fluoroadenosine, 2r,3r,4r,5r-5-2,6-diaminopurin-9-yl-4-fluoro-2-hydroxymethyl oxolan-3-ol, 2-amino-2/'-deoxy-2/'-fluoroadenosine, adenosine, 2-amino-2'-deoxy-2'-fluoro, 2,6-diamino-9-2-deoxy-2-fluoro-beta-d-ribofuranosyl-9h-purine, 1,2-dideoxy-1-2,6-diamino-9h-purin-9-yl-2-fluoro-beta-d-ribofuranose
(2R,3R,4R,5R)-5-(2,6-diaminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol

Specifications

White
134444-47-6
MFCD00900994
MHWHYOFBOWTAHZ-DXTOWSMRSA-N
(2R,3R,4R,5R)-5-(2,6-diaminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
9971101
Powder
2-Amino-2'-fluoro-2'-deoxyadenosine
250mg
C10H13FN6O3
2-amino-2'-deoxy-2'-fluoroadenosine, 2'-fluoro-2,6-diaminopurine-2'-deoxyriboside, 2-amino-2'-fluoro-2'-deoxyadenosine, 2'-amino-2'-deoxy-2-fluoroadenosine, 2r,3r,4r,5r-5-2,6-diaminopurin-9-yl-4-fluoro-2-hydroxymethyl oxolan-3-ol, 2-amino-2/'-deoxy-2/'-fluoroadenosine, adenosine, 2-amino-2'-deoxy-2'-fluoro, 2,6-diamino-9-2-deoxy-2-fluoro-beta-d-ribofuranosyl-9h-purine, 1,2-dideoxy-1-2,6-diamino-9h-purin-9-yl-2-fluoro-beta-d-ribofuranose
C1=NC2=C(N1C3C(C(C(O3)CO)O)F)N=C(N=C2N)N
284.251
284.25
99%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C