missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Amino-2'-deoxyinosine, 98%, Thermo Scientific Chemicals
$167.68 - $533.49
Chemical Identifiers
CAS | 75763-51-8 |
---|---|
Molecular Formula | C10H13N5O4 |
Molecular Weight (g/mol) | 267.25 |
MDL Number | MFCD09750831 |
InChI Key | PPLSURAOXNSSRH-GPGUAWMRNA-N |
Synonym | 2'-amino-2'-deoxyinosine, inosine,2'-amino-2'-deoxy-9ci, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, inosine, 2'-amino-2'-deoxy, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl purin-6-ol, 2?-amino-2?-deoxyinosine, 2'-deoxy-2'-amino-inosine, 2'-amino-2'-deoxyinosine,2'-nh2-di, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one |
PubChem CID | 195857 |
SMILES | N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=CNC2=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ65374MD
|
Thermo Scientific Chemicals
J65374MD |
250 mg |
N/A
|
|
|||||
AAJ6537403
|
Thermo Scientific Chemicals
J6537403 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75763-51-8 | |
267.25 | |
PPLSURAOXNSSRH-GPGUAWMRNA-N | |
195857 |
C10H13N5O4 | |
MFCD09750831 | |
2'-amino-2'-deoxyinosine, inosine,2'-amino-2'-deoxy-9ci, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, inosine, 2'-amino-2'-deoxy, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl purin-6-ol, 2?-amino-2?-deoxyinosine, 2'-deoxy-2'-amino-inosine, 2'-amino-2'-deoxyinosine,2'-nh2-di, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one | |
N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=CNC2=O |
Specifications
75763-51-8 | |
C10H13N5O4 | |
250 mg | |
PPLSURAOXNSSRH-GPGUAWMRNA-N | |
9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one | |
195857 | |
98% | |
2'-Amino-2'-deoxyinosine |
White to Yellow | |
MFCD09750831 | |
2'-amino-2'-deoxyinosine, inosine,2'-amino-2'-deoxy-9ci, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, inosine, 2'-amino-2'-deoxy, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl purin-6-ol, 2?-amino-2?-deoxyinosine, 2'-deoxy-2'-amino-inosine, 2'-amino-2'-deoxyinosine,2'-nh2-di, 9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one | |
N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=CNC2=O | |
267.25 | |
267.24 | |
Powder |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only