Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-2'-chloro-5-nitrobenzophenone, 98%

Chemical Identifiers

Chemical Name or Material 2-Amino-2'-chloro-5-nitrobenzophenone
CAS 2011-66-7
Formula Weight 276.68
MDL Number MFCD00792453
Beilstein 2814216
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6644006

  SDS
alfa aesar™
H6644006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6644014

  SDS
alfa aesar™
H6644014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6644022

  SDS
alfa aesar™
H6644022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 119° to 123°C
Formula Weight 276.68
Chemical Name or Material 2-Amino-2'-chloro-5-nitrobenzophenone
Physical Form Powder
Assay 98%
MDL Number MFCD00792453
CAS 2011-66-7
Color Pale yellow to yellow
Quantity 5g
Sensitivity Air Sensitive
Beilstein 2814216
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C