Please login to your online account to display your discounted pricing

2-Amino-1-propene-1,1,3-tricarbonitrile, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-1-propene-1, 1, 3-tricarbonitrile
Name Note 97%
CAS 868-54-2
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6 H4 N4
Linear Formula NCCH2C(NH2)=C(CN)2
Formula Weight 132.12
MDL Number MFCD00001851
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC148190050

  SDS
thermo fisher scientific
AC148190050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 132.12
Quantity 5g
Merck Index 15, 476
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Synonym 2-Amino-1, 1, 3-tricyanopropene
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula NCCH2C(NH2)=C(CN)2
Name Note 97%
CAS 868-54-2
Molecular Formula C6 H4 N4
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Assay Percent Range 97%
Chemical Name or Material 2-Amino-1-propene-1, 1, 3-tricarbonitrile
MDL Number MFCD00001851
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.