Learn More
(+/-)-2-Amino-1-propanol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0360814
Description
DL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.
Solubility
Fully miscible in water.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
Specifications
(+/-)-2-Amino-1-propanol | |
8°C | |
174°C to 176°C | |
C3H9NO | |
MFCD00008085 | |
UN2735 | |
Air sensitive; Hygroscopic | |
Fully miscible in water. | |
CC(N)CO | |
75.11 | |
75.11 |
6168-72-5 | |
0.965 | |
83°C (181°F) | |
1.4495 | |
25 g | |
1209234 | |
dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine | |
BKMMTJMQCTUHRP-UHFFFAOYNA-N | |
2-aminopropan-1-ol | |
5126 | |
98% |
Chemical Identifiers
6168-72-5 | |
75.11 | |
BKMMTJMQCTUHRP-UHFFFAOYNA-N | |
5126 | |
CC(N)CO |
C3H9NO | |
MFCD00008085 | |
dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine | |
2-aminopropan-1-ol |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 228-207-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only