missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-2-Amino-1-phenylethanol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 104320500

 View more versions of this product

Catalog No. AC104320500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
137.18
Clear yellow
100.0
H2NCH2CH(C6H5)OH
MFCD00008137
1.5580 to 1.5600 (20°C, 589nm)
Glass bottle
7568-93-6
C8H11NO
Viscous Liquid
50g
15, 7399
Authentic
98%
97.5
α-(Aminomethyl)benzyl alcohol
(±)-2-Amino-1-phenylethanol
98%

Chemical Identifiers

7568-93-6
100.0
C8H11NO
137.18
α-(Aminomethyl)benzyl alcohol
≥97.5%
97.5
98%
H2NCH2CH(C6H5)OH
MFCD00008137
(±)-2-Amino-1-phenylethanol, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger