missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-2-Amino-1-butanol, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 103251000

 View more versions of this product

Catalog No. AC103251000



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

(±)-2-Amino-1-butanol
96-20-8
100.0
Undesignated
176°C to 178°C
96% min. (GC)
Authentic
MFCD00008095
15, 422
100mL
Solubility in water: soluble. Other solubilities: soluble in ethanol and ether
JCBPETKZIGVZRE-UHFFFAOYSA-N
2-aminobutan-1-ol
22129
Liquid
0.5% max.
97%
96.0
0.9440g/mL
-2°C
84°C
C4H11NO
C2H5CH(NH2)CH2OH
Glass bottle
1.4508 to 1.4528
1-butanol, 2-amino, 1-hydroxy-2-butylamine, 1-hydroxymethyl propylamine, 2-amino-1-butanol, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol, 2-aminobutanol, 2-aminobutyl alcohol, butanol-2-amine, dl-2-amino-1-butanol
0.944
CCC(CO)N
89.14
89.14
97%

Chemical Identifiers

96-20-8
89.14
JCBPETKZIGVZRE-UHFFFAOYSA-N
22129
CCC(CO)N
C4H11NO
MFCD00008095
1-butanol, 2-amino, 1-hydroxy-2-butylamine, 1-hydroxymethyl propylamine, 2-amino-1-butanol, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol, 2-aminobutanol, 2-aminobutyl alcohol, butanol-2-amine, dl-2-amino-1-butanol
2-aminobutan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-488-2

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only