Please login to your online account to display your discounted pricing

(±)-2-Amino-1-butanol 97%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103255000

 View more versions of this product

Catalog No. AC103255000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 89.14
Quantity 500mL
Color Clear
Merck Index 15, 422
EINECS Number 235-940-2
CAS Min % 96.0
CAS Max % 100.0
Refractive Index 1.4508 to 1.4528 (20°C, 589nm)
Water 0.5% max.
Name Note 97%
Linear Formula C2H5CH(NH2)CH2OH
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material (±)-2-Amino-1-butanol
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C4H11NO
MDL Number MFCD00008095
CAS 96-20-8
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material (±)-2-Amino-1-butanol
Name Note 97%
CAS 96-20-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H11NO
Linear Formula C2H5CH(NH2)CH2OH
Formula Weight 89.14
MDL Number MFCD00008095
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents