missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-1-benzylbenzimidazole, 99+%, Thermo Scientific™

Chemical Identifiers

CAS 43182-10-1
Molecular Formula C14H13N3
Molecular Weight (g/mol) 223.28
InChI Key LXIDSOCBAAMGJX-UHFFFAOYSA-N
Synonym 1-benzyl-1,3-benzodiazol-2-amine, 1-benzyl-1h-benzimidazol-2-amine, 1-benzyl-1h-benzo d imidazol-2-amine, 1-benzyl-1h-benzoimidazol-2-ylamine, 1-benzyl-2-aminobenzimidazole, 1-benzylbenzimidazole-2-ylamine, 1h-benzimidazol-2-amine,1-phenylmethyl, 2-amino-1-benzylbenzimidazole, 2-amino-1-phenylmethyl-1h-benzimidazole, cbmicro_006128
PubChem CID 170742
IUPAC Name 1-benzylbenzimidazol-2-amine
SMILES C1=CC=C(C=C1)CN2C3=CC=CC=C3N=C2N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC276290010
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC276290010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

43182-10-1
223.28
1-benzyl-1,3-benzodiazol-2-amine, 1-benzyl-1h-benzimidazol-2-amine, 1-benzyl-1h-benzo d imidazol-2-amine, 1-benzyl-1h-benzoimidazol-2-ylamine, 1-benzyl-2-aminobenzimidazole, 1-benzylbenzimidazole-2-ylamine, 1h-benzimidazol-2-amine,1-phenylmethyl, 2-amino-1-benzylbenzimidazole, 2-amino-1-phenylmethyl-1h-benzimidazole, cbmicro_006128
1-benzylbenzimidazol-2-amine
C14H13N3
LXIDSOCBAAMGJX-UHFFFAOYSA-N
170742
C1=CC=C(C=C1)CN2C3=CC=CC=C3N=C2N

Specifications

43182-10-1
100.0
195°C to 198°C
Authentic
Glass bottle
1g
C1=CC=C(C=C1)CN2C3=CC=CC=C3N=C2N
223.28
223.28
99+%
99.0
White
99+%
C14H13N3
1-benzyl-1,3-benzodiazol-2-amine, 1-benzyl-1h-benzimidazol-2-amine, 1-benzyl-1h-benzo d imidazol-2-amine, 1-benzyl-1h-benzoimidazol-2-ylamine, 1-benzyl-2-aminobenzimidazole, 1-benzylbenzimidazole-2-ylamine, 1h-benzimidazol-2-amine,1-phenylmethyl, 2-amino-1-benzylbenzimidazole, 2-amino-1-phenylmethyl-1h-benzimidazole, cbmicro_006128
LXIDSOCBAAMGJX-UHFFFAOYSA-N
1-benzylbenzimidazol-2-amine
170742
Fine Crystalline Powder
2-Amino-1-benzylbenzimidazole, 99+%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 256-130-5

TSCA : TSCA

SDS
Product Certifications

RUO – Research Use Only