missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-1,3-propanediol hydrochloride, 98%

$296.19 - $296.19

Chemical Identifiers

CAS 73708-65-3
Molecular Formula C3H9NO2·HCl
Formula Weight 127.57
MDL Number MFCD00012592
Synonym Serinol hydrochloride
Chemical Name or Material 2-Amino-1,3-propanediol hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2541903 SDS Alfa Aesar™
B2541903
1g Each for $296.19
AAB2541901 SDS Alfa Aesar™
B2541901
0.25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

73708-65-3
127.57
Serinol hydrochloride
C3H9NO2·HCl
MFCD00012592
2-Amino-1,3-propanediol hydrochloride

Specifications

73708-65-3
1g
127.57
Serinol hydrochloride
98%
104° to 106°C
C3H9NO2·HCl
MFCD00012592
2-Amino-1,3-propanediol hydrochloride

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning