missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-1,3-propanediol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 401030010

 View more versions of this product

Catalog No. AC401030010


 

Specifications Description & Specifications

Specifications

≥97.5%
Adhering Crystalline Powder
White to off-white
100.0
Authentic
C3H9NO2
534-03-2
MFCD00051487
Glass bottle
91.11
1g
52° to 56°C
2-Amino-1, 3-propanediol
NH2CH(CH2OH)2
98%
97.5
Serinol

Chemical Identifiers

534-03-2
100.0
C3H9NO2
91.11
Serinol
≥97.5%
97.5
98%
NH2CH(CH2OH)2
MFCD00051487
2-Amino-1, 3-propanediol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger