Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-1,3,5-triazine-4,6-dithiol, tech. 90%

$91.46 - $91.46

Chemical Identifiers

Chemical Name or Material 2-Amino-1,3,5-triazine-4,6-dithiol
CAS 2770-75-4
Purity Grade Technical Grade
Molecular Formula C3H4N4S2
Formula Weight 160.22
MDL Number MFCD00047346
Synonym 2-Amino-4,6-dimercapto-s-triazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2051006

  SDS
alfa aesar™
B2051006
5g Each for $91.46
Description & Specifications

Specifications

Formula Weight 160.22
Synonym 2-Amino-4,6-dimercapto-s-triazine
CAS 2770-75-4
Melting Point >360°C
Assay 90%
MDL Number MFCD00047346
Purity Grade Technical Grade
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Quantity 5g
Molecular Formula C3H4N4S2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Amino-1,3,5-triazine-4,6-dithiol
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.