missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-1-(2-fluorophenyl)ethanol, 99%

$114.30 - $397.92

Chemical Identifiers

CAS 3225-74-9
Molecular Formula C8H10FNO
Formula Weight 155.17
MDL Number MFCD08060976
Chemical Name or Material 2-Amino-1-(2-fluorophenyl)ethanol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5077003 SDS Alfa Aesar™
H5077003
1g Each for $397.92
AAH50770MD SDS Alfa Aesar™
H50770MD
250mg Each for $114.30
Specifications

Description

Applications
2-Amino-1-(2-fluorophenyl)ethanol is used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

3225-74-9
155.17
2-Amino-1-(2-fluorophenyl)ethanol
C8H10FNO
MFCD08060976

Specifications

3225-74-9
155.17
1g
99%
C8H10FNO
MFCD08060976
2-Amino-1-(2-fluorophenyl)ethanol

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning