missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amidinopyridine hydrochloride, 97%

$338.64 - $1,419.22

Chemical Identifiers

CAS 51285-26-8
Molecular Formula C6H7N3·HCl
Formula Weight 157.6
MDL Number MFCD00052271
Beilstein 3562671
Synonym Pyridine-2-carboximidamide hydrochloride
Chemical Name or Material 2-Amidinopyridine hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1094206 SDS Alfa Aesar™
A1094206
5g Each for $338.64
AAA1094214 SDS Alfa Aesar™
A1094214
25g Each for $1,419.22
AAA1094203 SDS Alfa Aesar™
A1094203
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

51285-26-8
157.6
3562671
2-Amidinopyridine hydrochloride
C6H7N3·HCl
MFCD00052271
Pyridine-2-carboximidamide hydrochloride

Specifications

51285-26-8
157.6
3562671
Pyridine-2-carboximidamide hydrochloride
2-Amidinopyridine hydrochloride
C6H7N3·HCl
MFCD00052271
148° to 152°C
5g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning