Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Allylphenoxy)trimethylsilane, 98%

$70.50 - $886.62

Chemical Identifiers

Chemical Name or Material (2-Allylphenoxy)trimethylsilane
CAS 18042-43-8
Molecular Formula C12H18OSi
Formula Weight 206.36
MDL Number MFCD12407078
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3090506

 
alfa aesar™
H3090506
5g Each for $70.50

AAH3090514

 
alfa aesar™
H3090514
25g Each for $280.15

AAH3090522

 
alfa aesar™
H3090522
100g Each for $886.62
Description & Specifications

Specifications

Formula Weight 206.36
Chemical Name or Material (2-Allylphenoxy)trimethylsilane
Assay 98%
Molecular Formula C12H18OSi
MDL Number MFCD12407078
CAS 18042-43-8
Boiling Point 85°C (5mmHg)
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.