Learn More
2-Allyloxyethanol, 98%, Thermo Scientific™
Supplier: Thermo Fisher Scientific 449030010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Allyloxyethanol | |
0.9550g/mL | |
66°C | |
C5H10O2 | |
MFCD00020607 | |
1L | |
Solubility in water: soluble | |
GCYHRYNSUGLLMA-UHFFFAOYSA-N | |
2-prop-2-enoxyethanol | |
8116 | |
98% |
111-45-5 | |
159.0°C | |
97.5% min. (GC) | |
CH2CHCH2OCH2CH2OH | |
Glass bottle | |
2-2-propenyloxy ethanol, 2-alloxyethanol, 2-allyloxy ethanol, 2-allyloxyethanol, allyl 2-hydroxyethyl ether, allyl cellosolve, allyl glycol, ethanol, 2-2-propenyloxy, ethylene glycol monoallyl ether, usaf do-47 | |
0.955 | |
OCCOCC=C | |
102.13 | |
102.13 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 203-871-7
RUO – Research Use Only