Please login to your online account to display your discounted pricing

2-Allyloxyethanol, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Allyloxyethanol
Assay Percent Range 98%
Molecular Formula C5H10O2
Formula Weight 102.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC449032500

  SDS
thermo fisher scientific
AC449032500
Glass bottle 250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC449030010

  SDS
thermo fisher scientific
AC449030010
Glass bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 102.13
Quantity 250mL
Chemical Name or Material 2-Allyloxyethanol
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C5H10O2
CAS 111-45-5