missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Allyloxybenzaldehyde 90%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC380830050

 View more versions of this product

Catalog No. AC380830050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Allyloxybenzaldehyde
88.0
Authentic
1.5540 to 1.5560 (20°C, 589nm)
Liquid
90%
CH2CHCH2OC6H4CHO
162.19
Clear yellow to orange
28752-82-1
100.0
Glass bottle
Product may darken during storage
≥88%
C10 H10 O2
1.0790g/mL
MFCD00014130
5mL

Chemical Identifiers

28752-82-1
100.0
C10 H10 O2
162.19
2-Allyloxybenzaldehyde
88.0
90%
CH2CHCH2OC6H4CHO
MFCD00014130
≥88%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.