missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Allyloxybenzaldehyde 90%, ACROS Organics™

Chemical Identifiers

CAS 28752-82-1
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C10 H10 O2
Linear Formula CH2CHCH2OC6H4CHO
Formula Weight 162.19
MDL Number MFCD00014130
Chemical Name or Material 2-Allyloxybenzaldehyde
Assay ≥88%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC380830050 SDS Thermo Fisher Scientific
AC380830050
Glass bottle 5mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

28752-82-1
100.0
C10 H10 O2
162.19
2-Allyloxybenzaldehyde
88.0
90%
CH2CHCH2OC6H4CHO
MFCD00014130
≥88%

Specifications

162.19
5mL
1.0790g/mL
100.0
CH2CHCH2OC6H4CHO
C10 H10 O2
28752-82-1
Product may darken during storage
2-Allyloxybenzaldehyde
Liquid
Clear yellow to orange
88.0
Glass bottle
MFCD00014130
90%
1.5540 to 1.5560 (20°C, 589nm)
Authentic
≥88%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.