missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Allyl-N-Fmoc-DL-glycine, 98%

$211.27 - $845.08

Chemical Identifiers

CAS 221884-63-5
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.375
MDL Number MFCD02259489
InChI Key YVBLQCANYSFEBN-UHFFFAOYSA-N
Synonym fmoc-alpha-allyl-dl-glycine, fmoc-dl-allylglycine, 2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, s-n-fmoc-allyl-glycine, fmocdlallylglycine, fmoc-allyl-dl-gly-oh, fmoc-alpha-allyl-dl-gly-oh, 2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2-allyl-n-fmoc-dl-glycine, fmoc-2-amino-4-pentenoic acid;fmoc-allylglycine
PubChem CID 4052587
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid
SMILES C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6634506 View Documents Alfa Aesar™
H6634506
5g Each for $211.27
 
AAH6634514 View Documents Alfa Aesar™
H6634514
25g Each for $845.08
 
Specifications

Chemical Identifiers

221884-63-5
337.375
YVBLQCANYSFEBN-UHFFFAOYSA-N
4052587
C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C20H19NO4
MFCD02259489
fmoc-alpha-allyl-dl-glycine, fmoc-dl-allylglycine, 2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, s-n-fmoc-allyl-glycine, fmocdlallylglycine, fmoc-allyl-dl-gly-oh, fmoc-alpha-allyl-dl-gly-oh, 2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2-allyl-n-fmoc-dl-glycine, fmoc-2-amino-4-pentenoic acid;fmoc-allylglycine
2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid

Specifications

221884-63-5
MFCD02259489
YVBLQCANYSFEBN-UHFFFAOYSA-N
2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid
4052587
Solid
5g
C20H19NO4
fmoc-alpha-allyl-dl-glycine, fmoc-dl-allylglycine, 2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, s-n-fmoc-allyl-glycine, fmocdlallylglycine, fmoc-allyl-dl-gly-oh, fmoc-alpha-allyl-dl-gly-oh, 2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2-allyl-n-fmoc-dl-glycine, fmoc-2-amino-4-pentenoic acid;fmoc-allylglycine
C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
337.375
337.37
98%
2-Allyl-N-Fmoc-DL-glycine
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C