Please login to your online account to display your discounted pricing

2-Acrylamido-2-methylpropanesulfonic acid, 97%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC400292500

 View more versions of this product

Catalog No. AC400292500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder, Chunks or Chips
Formula Weight 207.25
Quantity 250g
Color White
Density 160g/mL
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula CH2CHCONHC(CH3)2CH2SO3H
CAS 15214-89-8
Molecular Formula C7H13NO4S
Assay Percent Range 97%
Chemical Name or Material 2-Acrylamido-2-methylpropanesulfonic acid
MDL Number MFCD00007522
Synonym 2-Methyl-2-[(1-oxo-2-propenyl)amino]-1-propanesulfonic acid
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Causes serious eye damage.

Specifications

Chemical Name or Material 2-Acrylamido-2-methylpropanesulfonic acid
CAS 15214-89-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H13NO4S
Linear Formula CH2CHCONHC(CH3)2CH2SO3H
Formula Weight 207.25
MDL Number MFCD00007522
Synonym 2-Methyl-2-[(1-oxo-2-propenyl)amino]-1-propanesulfonic acid
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Causes serious eye damage.

SDS & Safety Documents