Please login to your online account to display your discounted pricing

2-Acetylthiazole, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  265410250

 View more versions of this product

Catalog No. AC265410250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5450 to 1.5500 (20°C, 589nm)
Density 1.2200g/mL
Color White to slightly yellow
Quantity 25g
Chemical Name or Material 2-Acetylthiazole
CAS 24295-03-2
CAS Min % 98.5
CAS Max % 100.0
Physical Form White to slightly yellow powder or crystals
Assay 98.5 % min. (GC)
Assay Percent Range 99%
Molecular Formula C5H5NOS
Formula Weight 127.17g/mol
MDL Number MFCD00005324
Beilstein 27, 2617
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Acetylthiazole
CAS 24295-03-2
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5 % min. (GC)
Assay Percent Range 99%
Molecular Formula C5H5NOS
Formula Weight 127.17g/mol
MDL Number MFCD00005324
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents