Please login to your online account to display your discounted pricing

2-Acetylthiazole, 99%, ACROS Organics™

$179.52 - $179.52

Chemical Identifiers

Chemical Name or Material 2-Acetylthiazole
CAS 24295-03-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C5H5NOS
Formula Weight 127.17
MDL Number MFCD00005324
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC265410250

  SDS
thermo fisher scientific
AC265410250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC265410050

  SDS
thermo fisher scientific
265410050
Glass bottle 5g Each for $179.52
Description & Specifications

Specifications

Physical Form Powder or Crystals
Assay ≥98.5% (GC)
Formula Weight 127.17
Quantity 25g
Color White to slightly yellow
Density 1.2200g/mL
Beilstein 27, 2617
CAS Max % 100.0
Chemical Name or Material 2-Acetylthiazole
Assay Percent Range 99%
CAS 24295-03-2
Infrared Spectrum Authentic
Molecular Formula C5H5NOS
Packaging Glass bottle
MDL Number MFCD00005324
Refractive Index 1.5450 to 1.5500 (20°C, 589nm)
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.