Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetylquinoxaline, 97%

$98.66 - $246.28

Chemical Identifiers

Chemical Name or Material 2-Acetylquinoxaline
CAS 25594-62-1
Molecular Formula C10H8N2O
Formula Weight 172.19
MDL Number MFCD07808559
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2668003

 
alfa aesar™
H2668003
1g Each for $246.28

AAH26680MD

 
alfa aesar™
H26680MD
250mg Each for $98.66
Description & Specifications

Specifications

Formula Weight 172.19
Chemical Name or Material 2-Acetylquinoxaline
MDL Number MFCD07808559
CAS 25594-62-1
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 97%
Molecular Formula C10H8N2O
Health Hazard 1 Warning
Melting Point 76° to 79°C

Applications
2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.