Learn More
2-Acetylpyrrole, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC252490250
Specifications
2-Acetylpyrrole | |
Glass bottle | |
White to Beige | |
25g | |
98.5 | |
98.5% min. (GC) | |
MFCD00005220 | |
1-1h-pyrrol-2-yl ethan-1-one, 1-1h-pyrrol-2-yl ethanone, 2-acetyl pyrrole, 2-acetyl-1h-pyrrole, 2-acetylpyrrole, 2-pyrrolyl methyl ketone, 2-pyrrolylethanone, ethanone, 1-1h-pyrrol-2-yl, ketone, methyl pyrrol-2-yl, methyl pyrrol-2-yl ketone | |
IGJQUJNPMOYEJY-UHFFFAOYSA-N | |
1-(1H-pyrrol-2-yl)ethanone | |
14079 | |
109.13 | |
99% |
Authentic | |
220°C | |
88°C to 91°C | |
1072-83-9 | |
100.0 | |
C6H7NO | |
21, 271 | |
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents,soluble in ethanol and ether (20°C),soluble in alcohol and propylene glycol | |
CC(=O)C1=CC=CN1 | |
109.13 | |
CHEBI:59981 | |
Crystalline Powder |
Chemical Identifiers
1072-83-9 | |
109.13 | |
IGJQUJNPMOYEJY-UHFFFAOYSA-N | |
14079 | |
1-(1H-pyrrol-2-yl)ethanone |
C6H7NO | |
MFCD00005220 | |
1-1h-pyrrol-2-yl ethan-1-one, 1-1h-pyrrol-2-yl ethanone, 2-acetyl pyrrole, 2-acetyl-1h-pyrrole, 2-acetylpyrrole, 2-pyrrolyl methyl ketone, 2-pyrrolylethanone, ethanone, 1-1h-pyrrol-2-yl, ketone, methyl pyrrol-2-yl, methyl pyrrol-2-yl ketone | |
CHEBI:59981 | |
CC(=O)C1=CC=CN1 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 214-016-2