missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetylpyrrole, 98%

Manufacturer:  Alfa Aesar™ A1459303

 View more versions of this product

Catalog No. AAA1459303



Specifications

Specifications

2-Acetylpyrrole
220°C
1g
C6H7NO
1882
IGJQUJNPMOYEJY-UHFFFAOYSA-N
1-(1H-pyrrol-2-yl)ethanone
14079
109.13
Characteristic
88°C to 92°C
1072-83-9
MFCD00005220
2-acetylpyrrole, 1-1h-pyrrol-2-yl ethanone, 1-1h-pyrrol-2-yl ethan-1-one, 2-acetyl pyrrole, 2-pyrrolylethanone, 2-acetyl-1h-pyrrole, methyl pyrrol-2-yl ketone, ethanone, 1-1h-pyrrol-2-yl, 2-pyrrolyl methyl ketone, ketone, methyl pyrrol-2-yl
CC(=O)C1=CC=CN1
109.128
CHEBI:59981
98%

Chemical Identifiers

1072-83-9
109.128
IGJQUJNPMOYEJY-UHFFFAOYSA-N
14079
1-(1H-pyrrol-2-yl)ethanone
C6H7NO
MFCD00005220
2-acetylpyrrole, 1-1h-pyrrol-2-yl ethanone, 1-1h-pyrrol-2-yl ethan-1-one, 2-acetyl pyrrole, 2-pyrrolylethanone, 2-acetyl-1h-pyrrole, methyl pyrrol-2-yl ketone, ethanone, 1-1h-pyrrol-2-yl, 2-pyrrolyl methyl ketone, ketone, methyl pyrrol-2-yl
CHEBI:59981
CC(=O)C1=CC=CN1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OB5970000

TSCA : TSCA

Documents