Learn More
Alfa Aesar™ 2-Acetylpyrrole, 98%
Manufacturer: Alfa Aesar™ A1459314
Specifications
2-Acetylpyrrole | |
220°C | |
25g | |
C6H7NO | |
1882 | |
IGJQUJNPMOYEJY-UHFFFAOYSA-N | |
1-(1H-pyrrol-2-yl)ethanone | |
14079 | |
109.13 |
Characteristic | |
88°C to 92°C | |
1072-83-9 | |
MFCD00005220 | |
2-acetylpyrrole, 1-1h-pyrrol-2-yl ethanone, 1-1h-pyrrol-2-yl ethan-1-one, 2-acetyl pyrrole, 2-pyrrolylethanone, 2-acetyl-1h-pyrrole, methyl pyrrol-2-yl ketone, ethanone, 1-1h-pyrrol-2-yl, 2-pyrrolyl methyl ketone, ketone, methyl pyrrol-2-yl | |
CC(=O)C1=CC=CN1 | |
109.128 | |
CHEBI:59981 | |
98% |
Chemical Identifiers
1072-83-9 | |
109.128 | |
IGJQUJNPMOYEJY-UHFFFAOYSA-N | |
14079 | |
1-(1H-pyrrol-2-yl)ethanone |
C6H7NO | |
MFCD00005220 | |
2-acetylpyrrole, 1-1h-pyrrol-2-yl ethanone, 1-1h-pyrrol-2-yl ethan-1-one, 2-acetyl pyrrole, 2-pyrrolylethanone, 2-acetyl-1h-pyrrole, methyl pyrrol-2-yl ketone, ethanone, 1-1h-pyrrol-2-yl, 2-pyrrolyl methyl ketone, ketone, methyl pyrrol-2-yl | |
CHEBI:59981 | |
CC(=O)C1=CC=CN1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : OB5970000
TSCA : TSCA