missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetylnaphthalene, 99%

Manufacturer:  Alfa Aesar™ A1479330

 View more versions of this product

Catalog No. AAA1479330



Description

Description

Applications
2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.

Solubility
Soluble in ethanol, acetone and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications

Specifications

2-Acetylnaphthalene
300°C to 301°C
250g
C12H10O
UN3077
2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
CC(=O)C1=CC2=CC=CC=C2C=C1
170.211
CHEBI:52364
99%
168°C (334°F)
52°C to 56°C
93-08-3
MFCD00004108
774965
XSAYZAUNJMRRIR-UHFFFAOYSA-N
1-naphthalen-2-ylethanone
7122
170.21

Chemical Identifiers

93-08-3
170.211
XSAYZAUNJMRRIR-UHFFFAOYSA-N
7122
1-naphthalen-2-ylethanone
C12H10O
MFCD00004108
2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
CHEBI:52364
CC(=O)C1=CC2=CC=CC=C2C=C1
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : AL2988000

TSCA : TSCA

SDS Documents