missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetylnaphthalene, 99%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific A147930B

Catalog No. AAA147930B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Acetylnaphthalene is used as a flavoring agent in food. It serves as an intermediate in the preparation of piperidinyl pyrazoles as potent DNA gyrase inhibitors. Further, it is used in air care products, cleaning and furnishing care products. In addition to this, it is used in the synthesis of aromatic enone and dienone analogues of curcumin as angiogenesis inhibitors.

Solubility
Soluble in ethanol, acetone and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications

Specifications

2-Acetylnaphthalene
52°C to 56°C
168°C (334°F)
MFCD00004108
774965
1-2-naphthyl ethanone, 1-naphthalen-2-yl ethanone, 2'-acetonaphthone, 2-acetonaphthone, 2-acetylnaphthalene, 2-naphthyl methyl ketone, acetonaphthone, ethanone, 1-2-naphthalenyl, methyl 2-naphthyl ketone, oranger cyrstals
XSAYZAUNJMRRIR-UHFFFAOYSA-N
1-naphthalen-2-ylethanone
7122
170.21
93-08-3
300°C to 301°C
C12H10O
UN3077
1000g
Soluble in ethanol,acetone and ether. Insoluble in water.
CC(=O)C1=CC2=CC=CC=C2C=C1
170.211
CHEBI:52364
99%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 202-216-2

RTECSNumber : AL2988000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only