Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetylfluorene, 98%

$442.99 - $442.99

Chemical Identifiers

Chemical Name or Material 2-Acetylfluorene
CAS 781-73-7
Molecular Formula C15H12O
Formula Weight 208.26
MDL Number MFCD00001124
Beilstein 1871711
Synonym 2-Fluorenyl methyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2284022

  SDS
alfa aesar™
B2284022
100g Each for $442.99
Description & Specifications

Specifications

Formula Weight 208.26
Synonym 2-Fluorenyl methyl ketone
Flash Point 217°C (422°F)
Melting Point 127° to 132°C
MDL Number MFCD00001124
Odor Odorless
RTECS Number OB3800000
CAS 781-73-7
Molecular Formula C15H12O
Beilstein 1871711
Quantity 100g
Chemical Name or Material 2-Acetylfluorene
Assay 98%