Please login to your online account to display your discounted pricing

2-Acetylcyclopentanone, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Acetylcyclopentanone
CAS 1670-46-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H10O2
Linear Formula CH3COC5H7(=O)
Formula Weight 126.15
MDL Number MFCD00001413
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC177031000

  SDS
thermo fisher scientific
AC177031000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC177030050

  SDS
thermo fisher scientific
177030050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 126.15
Quantity 100g
Color Clear light yellow to brown
Density 1.0400g/mL
Beilstein 07, 558
CAS Max % 100.0
Chemical Name or Material 2-Acetylcyclopentanone
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
Linear Formula CH3COC5H7(=O)
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H10O2
MDL Number MFCD00001413
CAS 1670-46-8
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Safety and Handling

Warning

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.