Please login to your online account to display your discounted pricing

2-Acetylcyclopentanone, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  177030050

 View more versions of this product

Catalog No. AC177030050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 126.15
Quantity 5g
Color Clear light yellow to brown
Beilstein 07, 558
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
Linear Formula CH3COC5H7(=O)
Assay Percent Range 99%
CAS 1670-46-8
Molecular Formula C7H10O2
Chemical Name or Material 2-Acetylcyclopentanone
Health Hazard 3 GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
CAS Min % 98.5
MDL Number MFCD00001413
Infrared Spectrum Authentic
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2-Acetylcyclopentanone
CAS 1670-46-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H10O2
Linear Formula CH3COC5H7(=O)
Formula Weight 126.15
MDL Number MFCD00001413
Accessories

Warning

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents