Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetylcyclohexanone, 98%

$44.45 - $108.04

Chemical Identifiers

Chemical Name or Material 2-Acetylcyclohexanone
CAS 874-23-7
Molecular Formula C8H12O2
Formula Weight 140.18
MDL Number MFCD00001633
Beilstein 1858621
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0275306

  SDS
alfa aesar™
L0275306
5g Each for $44.45

AAL0275314

  SDS
alfa aesar™
L0275314
25g Each for $108.04
Description & Specifications

Specifications

Formula Weight 140.18
Chemical Name or Material 2-Acetylcyclohexanone
CAS 874-23-7
Health Hazard 1 Warning
Beilstein 1858621
Molecular Formula C8H12O2
Assay 98%
Flash Point 79°C (174°F)
Quantity 5g
TSCA TSCA
Boiling Point 111° to 112°C (18mmHg)
MDL Number MFCD00001633

Applications
2-Acetylcyclohexanone is useful for synthesis of anilinoethanolamines, 1-(pyridyl)ethanol derivatives that can reduce keto-esters and ketones. It is also used in the synthesis of phenothiazine derivatives that can inhibit prote in farnesyltransferase molecules and some that show anti-proliferative activity.

Solubility
Not miscible in water.

Notes
Incompatible with Strong oxidizing agents, Strong bases. Air-sensitive.

Safety and Handling

Warning