missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetylcyclohexanone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 346190250

 View more versions of this product

Catalog No. AC346190250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
140.18
Clear colorless to light yellow
100.0
Glass bottle
874-23-7
97%
CH3COC6H9(=O)
1.5090 to 1.5110 (20°C, 589nm)
Liquid
25g
07, 559
96.0
C8H12O2
2-Acetylcyclohexanone
97%
MFCD00001633
Authentic

Chemical Identifiers

874-23-7
100.0
C8H12O2
140.18
2-Acetylcyclohexanone, 97%
96.0
97%
CH3COC6H9(=O)
MFCD00001633
≥96.0%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 212-858-5

TSCA : TSCA