missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetylbenzoic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1021618

 View more versions of this product

Catalog No. AAA1021618



Description

Description

2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.

Solubility
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.

Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Specifications

2-Acetylbenzoic acid
115°C to 118°C
C9H8O3
50 g
o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
QDAWXRKTSATEOP-UHFFFAOYSA-N
2-acetylbenzoic acid
68474
≥98%
577-56-0
Odorless
MFCD00002475
1210557
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.
CC(=O)C1=CC=CC=C1C(O)=O
164.16
164.16

Chemical Identifiers

577-56-0
164.16
QDAWXRKTSATEOP-UHFFFAOYSA-N
68474
CC(=O)C1=CC=CC=C1C(O)=O
C9H8O3
MFCD00002475
o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
2-acetylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-413-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only