Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetylanthracene, 97%

$238.07 - $1,033.64

Chemical Identifiers

Chemical Name or Material 2-Acetylanthracene
CAS 10210-32-9
Molecular Formula C16H12O
Formula Weight 220.27
MDL Number MFCD00156727
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4475106

  SDS
alfa aesar™
4475106
5g Each for $238.07

AA4475114

  SDS
alfa aesar™
4475114
25g Each for $1,033.64
Description & Specifications

Specifications

Formula Weight 220.27
Chemical Name or Material 2-Acetylanthracene
Physical Form Powder
Assay 97%
Molecular Formula C16H12O
MDL Number MFCD00156727
CAS 10210-32-9
Melting Point 188° to 192°C
Quantity 5g