Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-7-methoxybenzo[b]furan, 97%

$56.50 - $907.07

Chemical Identifiers

Chemical Name or Material 2-Acetyl-7-methoxybenzo[b]furan
CAS 43071-52-9
Molecular Formula C11H10O3
Formula Weight 190.2
MDL Number MFCD00016911
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2113903

  SDS
alfa aesar™
B2113903
1g Each for $56.50

AAB2113914

  SDS
alfa aesar™
B2113914
25g Each for $907.07
Description & Specifications

Specifications

Formula Weight 190.2
Assay 97%
Molecular Formula C11H10O3
MDL Number MFCD00016911
Quantity 1g
Melting Point 91° to 94°C
CAS 43071-52-9
Chemical Name or Material 2-Acetyl-7-methoxybenzo[b]furan