Learn More
2-Acetyl-6-methylpyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6599703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Acetyl-6-methylpyridine | |
Yellow | |
84°C (183°F) | |
1.515 | |
1 g | |
2-acetyl-6-methylpyridine, 1-6-methylpyridin-2-yl ethanone, 1-6-methylpyridin-2-yl ethan-1-one, ketone, methyl 6-methyl-2-pyridyl, 6-acetyl-2-picoline, 1-6-methyl-pyridin-2-yl-ethanone, 1-6-methyl-2-pyridinyl ethanone, 6-methyl-2-acetylpyridine, ethanone, 1-6-methyl-2-pyridinyl, 6-acetyl-2-methylpyridine | |
CC1=CC=CC(=N1)C(=O)C | |
135.166 | |
135.17 | |
Solid |
6940-57-4 | |
209°C | |
C8H9NO | |
MFCD06658293 | |
109564 | |
FPQMUQPPAYCAME-UHFFFAOYSA-N | |
1-(6-methylpyridin-2-yl)ethanone | |
138872 | |
98% |
Chemical Identifiers
6940-57-4 | |
135.166 | |
FPQMUQPPAYCAME-UHFFFAOYSA-N | |
138872 | |
CC1=CC=CC(=N1)C(=O)C |
C8H9NO | |
MFCD06658293 | |
2-acetyl-6-methylpyridine, 1-6-methylpyridin-2-yl ethanone, 1-6-methylpyridin-2-yl ethan-1-one, ketone, methyl 6-methyl-2-pyridyl, 6-acetyl-2-picoline, 1-6-methyl-pyridin-2-yl-ethanone, 1-6-methyl-2-pyridinyl ethanone, 6-methyl-2-acetylpyridine, ethanone, 1-6-methyl-2-pyridinyl, 6-acetyl-2-methylpyridine | |
1-(6-methylpyridin-2-yl)ethanone |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only