Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-6-methylpyridine, 98%

Chemical Identifiers

Chemical Name or Material 2-Acetyl-6-methylpyridine
CAS 6940-57-4
Formula Weight 135.17
MDL Number MFCD06658293
Beilstein 109564
Synonym 6-Acetyl-2-picoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6599703

  SDS
alfa aesar™
H6599703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6599706

  SDS
alfa aesar™
H6599706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65997MD

  SDS
alfa aesar™
H65997MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135.17
CAS 6940-57-4
Chemical Name or Material 2-Acetyl-6-methylpyridine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color Pale yellow
Storage Note 1 2° to 8°C
Synonym 6-Acetyl-2-picoline
MDL Number MFCD06658293
Beilstein 109564
Assay 98%
Health Hazard 1 Warning
Quantity 1g
Physical Form Solid
Boiling Point 209°C
Refractive Index 1.5150
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C