Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-6-methoxynaphthalene, 98%

$207.80 - $2,259.66

Chemical Identifiers

Chemical Name or Material 2-Acetyl-6-methoxynaphthalene
CAS 3900-45-6
Molecular Formula C13H12O2
Formula Weight 200.24
MDL Number MFCD00021643
Beilstein 1871098
Synonym 6-Methoxy-2'-acetonaphthone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1803114

 
alfa aesar™
A1803114
25g Each for $207.80

AAA1803122

 
alfa aesar™
A1803122
100g Each for $566.46

AAA1803136

 
alfa aesar™
A1803136
500g Each for $2,259.66
Description & Specifications

Specifications

Formula Weight 200.24
Chemical Name or Material 2-Acetyl-6-methoxynaphthalene
Assay 98%
Molecular Formula C13H12O2
MDL Number MFCD00021643
CAS 3900-45-6
Beilstein 1871098
Synonym 6-Methoxy-2'-acetonaphthone
Melting Point 107° to 109°C
Boiling Point 155° to 160°C (0.4mmHg)
Quantity 25g