missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Acetyl-6-methoxynaphthalene, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 134780010

Catalog No. AC134780010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Acetyl-6-methoxynaphthalene
3900-45-6
100.0
108.0°C to 109.0°C
C13H12O2
CH3OC10H6COCH3
Glass bottle
1-6-methoxy-2-naphthyl ethan-1-one, 1-6-methoxy-2-naphthyl ethanone, 1-6-methoxynaphthalen-2-yl ethanone, 2-acetyl-6-methoxy naphthalene, 2-acetyl-6-methoxynaphthalene, 6'-methoxy-2'-acetonaphthone, 6-methoxy-2-acetyl naphthalene, 6-methoxy-2-acetylnaphthalene, ethanone, 1-6-methoxy-2-naphthalenyl, unii-61vbh3537c
GGWCZBGAIGGTDA-UHFFFAOYSA-N
1-(6-methoxynaphthalen-2-yl)ethanone
77506
Powder
98%
97.5
Beige-Yellow
98%
Authentic
MFCD00021643
1g
Solubility in water: insoluble
CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC
200.24
200.24
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 223-453-8

TSCA : TSCA

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only