missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-6-bromopyridine, 97%

Manufacturer:  Alfa Aesar™ H55981MD

 View more versions of this product

Catalog No. AAH55981MD



Description

Description

Applications
2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2-Acetyl-6-bromopyridine
250mg
C7H6BrNO
1-6-bromo-2-pyridinyl-1-ethanone
CC(=O)C1=NC(=CC=C1)Br
200.035
200.03
51°C to 55°C
49669-13-8
MFCD00272200
RUJTWTUYVOEEFW-UHFFFAOYSA-N
1-(6-bromopyridin-2-yl)ethanone
11298578
97%

Chemical Identifiers

49669-13-8
200.035
RUJTWTUYVOEEFW-UHFFFAOYSA-N
11298578
CC(=O)C1=NC(=CC=C1)Br
C7H6BrNO
MFCD00272200
1-6-bromo-2-pyridinyl-1-ethanone
1-(6-bromopyridin-2-yl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents