Learn More
2-Acetyl-6-bromopyridine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5598106
Description
2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.
Solubility
Soluble in methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
2-Acetyl-6-bromopyridine | |
51°C to 55°C | |
MFCD00272200 | |
2-acetyl-6-bromopyridine, 2-bromo-6-acetylpyridine, 1-6-bromopyridin-2-yl ethanone, 1-6-bromopyridin-2-yl ethan-1-one, 1-6-bromo-2-pyridinyl-1-ethanone, ethanone, 1-6-bromo-2-pyridinyl, bromopyridinylethanone, pubchem13351, acmc-1ap21, ksc490s1f | |
RUJTWTUYVOEEFW-UHFFFAOYSA-N | |
1-(6-bromopyridin-2-yl)ethanone | |
11298578 | |
97% |
49669-13-8 | |
C7H6BrNO | |
5 g | |
Soluble in methanol. | |
CC(=O)C1=NC(=CC=C1)Br | |
200.035 | |
200.03 |
Chemical Identifiers
49669-13-8 | |
200.035 | |
RUJTWTUYVOEEFW-UHFFFAOYSA-N | |
11298578 | |
CC(=O)C1=NC(=CC=C1)Br |
C7H6BrNO | |
MFCD00272200 | |
2-acetyl-6-bromopyridine, 2-bromo-6-acetylpyridine, 1-6-bromopyridin-2-yl ethanone, 1-6-bromopyridin-2-yl ethan-1-one, 1-6-bromo-2-pyridinyl-1-ethanone, ethanone, 1-6-bromo-2-pyridinyl, bromopyridinylethanone, pubchem13351, acmc-1ap21, ksc490s1f | |
1-(6-bromopyridin-2-yl)ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only