missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-6-bromopyridine, 97%

Manufacturer:  Alfa Aesar™ H5598103

 View more versions of this product

Catalog No. AAH5598103



Description

Description

Applications
2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2-Acetyl-6-bromopyridine
1g
C7H6BrNO
1-6-bromo-2-pyridinyl-1-ethanone
RUJTWTUYVOEEFW-UHFFFAOYSA-N
1-(6-bromopyridin-2-yl)ethanone
11298578
97%
51°C to 55°C
49669-13-8
MFCD00272200
Soluble in methanol.
CC(=O)C1=NC(=CC=C1)Br
200.035
200.03

Chemical Identifiers

49669-13-8
200.035
RUJTWTUYVOEEFW-UHFFFAOYSA-N
11298578
CC(=O)C1=NC(=CC=C1)Br
C7H6BrNO
MFCD00272200
1-6-bromo-2-pyridinyl-1-ethanone
1-(6-bromopyridin-2-yl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents