Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Acetyl-5-nitrobenzo[b]furan, 98%

$111.95 - $111.95

Chemical Identifiers

Chemical Name or Material 2-Acetyl-5-nitrobenzo[b]furan
CAS 23136-39-2
Molecular Formula C10H7NO4
Formula Weight 205.18
MDL Number MFCD00051634
Beilstein 1684589
Synonym Methyl 5-nitrobenzo[b]furanyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0630803

  SDS
alfa aesar™
L0630803
1g Each for $111.95
Description & Specifications

Specifications

Formula Weight 205.18
Chemical Name or Material 2-Acetyl-5-nitrobenzo[b]furan
MDL Number MFCD00051634
CAS 23136-39-2
Molecular Formula C10H7NO4
Assay 98%
Synonym Methyl 5-nitrobenzo[b]furanyl ketone
Melting Point 173° to 177°C
Quantity 1g
Beilstein 1684589